MilliporeSigma
All Photos(1)

107107

Sigma-Aldrich

2,7-Dinitro-9-fluorenone

97%

Empirical Formula (Hill Notation):
C13H6N2O5
CAS Number:
Molecular Weight:
270.20
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

97%

form

solid

mp

292-295 °C (lit.)

solubility

DMF: soluble 25 mg/mL, clear (yellow to yellow-green)

SMILES string

[O-][N+](=O)c1ccc2-c3ccc(cc3C(=O)c2c1)[N+]([O-])=O

InChI

1S/C13H6N2O5/c16-13-11-5-7(14(17)18)1-3-9(11)10-4-2-8(15(19)20)6-12(10)13/h1-6H

InChI key

HDVGAFBXTXDYIB-UHFFFAOYSA-N

General description

2,7-Dinitro-9-fluorenone forms 1:1 Π-complex with tetrathiafulvalene.

Application

2,7-Dinitro-9-fluorenone was used to develop sensor for determination of submicromolar concentrations of selected genotoxic nitro compounds using mercury meniscus modified silver solid amalgam electrode coupled with direct current voltammetry or differential pulse voltammetry. It was used to study mechanism of generation of spontaneous and mutation-induced deletion revertants of hisG428 in Salmonella tester strain TA102 .

Packaging

10, 50 g in glass bottle

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service