Documents

10984

Sigma-Aldrich

Brominated Wang resin

extent of labeling: ~1.0 mmol/g loading

Synonym(s):
2-Bromopropionylpolystyrene, α-Bromopropiophenone, polymer-bound
MDL number:
Pricing and availability is not currently available.

extent of labeling

~1.0 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

70-90 mesh

Looking for similar products? Visit Product Comparison Guide

Other Notes

Photolabile polystyrene resin for the synthesis of protected peptides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Solid phase synthesis of protected peptides via photolytic cleavage of the alpha-methylphenacyl ester anchoring linkage.
S S Wang
The Journal of organic chemistry, 41(20), 3258-3261 (1976-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.