111392

Sigma-Aldrich

Vinylcyclopentane

99%

Synonym(s):
Ethenylcyclopentane, Cyclopentylethylene
Linear Formula:
C5H9CH=CH2
CAS Number:
Molecular Weight:
96.17
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

assay

99%

refractive index

n20/D 1.436 (lit.)

bp

97 °C (lit.)

density

0.704 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1CCCC1

InChI

1S/C7H12/c1-2-7-5-3-4-6-7/h2,7H,1,3-6H2

InChI key

BEFDCLMNVWHSGT-UHFFFAOYSA-N

Related Categories

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

F

Risk Statement

11

Safety Statement

16-29-33

RIDADR

UN 3295BF 3 / PGII

WGK Germany

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.