All Photos(1)

118052

Sigma-Aldrich

N-(4-Bromophenyl)-2-chloroacetamide

96%

Linear Formula:
ClCH2CONHC6H4Br
CAS Number:
Molecular Weight:
248.50
MDL number:
PubChem Substance ID:

assay

96%

mp

180-184 °C (dec.) (lit.)

SMILES string

ClCC(=O)Nc1ccc(Br)cc1

InChI

1S/C8H7BrClNO/c9-6-1-3-7(4-2-6)11-8(12)5-10/h1-4H,5H2,(H,11,12)

InChI key

FRZKCMCCQAJIBN-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service