MilliporeSigma
All Photos(1)

140058

Sigma-Aldrich

2,4-Dinitroanisole

98%

Linear Formula:
(O2N)2C6H3OCH3
CAS Number:
Molecular Weight:
198.13
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

94-96 °C (lit.)

density

1.34 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Christopher I Olivares et al.
Environmental science & technology, 51(22), 13327-13334 (2017-10-27)
2,4-Dinitrosanisole (DNAN) is an insensitive munitions component replacing conventional explosives. While DNAN is known to biotransform in soils to aromatic amines and azo-dimers, it is seldom mineralized by indigenous soil bacteria. Incorporation of DNAN biotransformation products into soil as humus-bound
Chunlei Wang et al.
Journal of hazardous materials, 396, 122627-122627 (2020-04-20)
The environmental fate of insensitive munitions compounds, such as 2,4-dinitroanisole (DNAN), has drawn increasing attention because of their growing use in military activities. One of the main attenuation mechanisms of DNAN degradation in aqueous environments is alkaline hydrolysis. We investigated
Raju Khatiwada et al.
Chemosphere, 201, 790-798 (2018-03-20)
Abiotic transformation of anthropogenic compounds by redox-active metal oxides affects contaminant fate in soil. The capacity of birnessite and ferrihydrite to oxidize the insensitive munitions compound, 2,4-dinitroanisol (DNAN), and its amine-containing daughter products, 2-methoxy-5-nitro aniline (MENA) and 2,4-diaminoanisole (DAAN), was
Christopher I Olivares et al.
Journal of hazardous materials, 304, 214-221 (2015-11-10)
Recent studies have begun to assess the environmental fate and toxicity of 2,4-dinitroanisole (DNAN), an insensitive munition compound of interest to defense agencies. Aerobic and anaerobic DNAN biotransformation in soils was evaluated in this study. Under aerobic conditions, there was
Matthew J Berens et al.
Environmental science. Processes & impacts, 21(1), 51-62 (2018-11-30)
The recent development of insensitive munitions, such as 2,4-dinitroanisole (DNAN), as components of military explosives has generated concern for potential subsurface contamination and created a need to fully characterize their transformation processes. Compound specific isotope analysis (CSIA) has proven to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service