149306

Sigma-Aldrich

Methoxyacetaldehyde diethyl acetal

97%

Linear Formula:
CH3OCH2CH(OC2H5)2
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.399 (lit.)

bp

144-148 °C (lit.)

density

0.911 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCOC(COC)OCC

InChI

1S/C7H16O3/c1-4-9-7(6-8-3)10-5-2/h7H,4-6H2,1-3H3

InChI key

PCYADPMXIILFTP-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3271 3 / PGIII

WGK Germany

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.