MilliporeSigma
All Photos(1)

154768

Millipore

Cyclopentane

≥75% cyclopentane basis, spectrophotometric grade

Empirical Formula (Hill Notation):
C5H10
CAS Number:
Molecular Weight:
70.13
Beilstein:
1900195
EC Number:
MDL number:
PubChem Substance ID:

grade

spectrophotometric grade

vapor density

~2 (vs air)

vapor pressure

18.93 psi

assay

≥75% cyclopentane basis

autoignition temp.

682 °F

expl. lim.

8.7 %

refractive index

n20/D 1.405 (lit.)

bp

50 °C (lit.)

mp

−94 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

SMILES string

C1CCCC1

InChI

1S/C5H10/c1-2-4-5-3-1/h1-5H2

InChI key

RGSFGYAAUTVSQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains C6 saturated hydrocarbons

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service