All Photos(1)

155446

Sigma-Aldrich

Benzyl diethyl phosphite

Linear Formula:
C6H5CH2OP(OC2H5)2
CAS Number:
Molecular Weight:
228.22
EC Number:
MDL number:
PubChem Substance ID:

refractive index

n20/D 1.493 (lit.)

bp

110 °C/2 mmHg (lit.)

density

1.076 g/mL at 25 °C (lit.)

SMILES string

CCOP(OCC)OCc1ccccc1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service