All Photos(1)

155624

Sigma-Aldrich

2′-(2-Chlorobenzoyl)-2,4′-dichloroacetanilide

98%

Synonym(s):
2-Chloroacetamido-2′,5-dichlorobenzophenone
Linear Formula:
ClCH2CONHC6H3(Cl)COC6H4Cl
CAS Number:
Molecular Weight:
342.60
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

159-161 °C (lit.)

SMILES string

ClCC(=O)Nc1ccc(Cl)cc1C(=O)c2ccccc2Cl

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service