All Photos(1)

164070

Sigma-Aldrich

trans-4-Octyl-α-chloro-4′-ethoxystilbene

≥99%

Linear Formula:
CH3(CH2)7C6H4C(Cl)=CHC6H4OC2H5
CAS Number:
Molecular Weight:
370.96
EC Number:
MDL number:

assay

≥99%

mp

32 °C (lit.)

SMILES string

CCCCCCCCc1ccc(cc1)\C(Cl)=C\c2ccc(OCC)cc2

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service