All Photos(1)

180319

Sigma-Aldrich

4-Cyanophenyl 4-heptylbenzoate

99%

Linear Formula:
CH3(CH2)6C6H4CO2C6H4CN
CAS Number:
Molecular Weight:
321.41
EC Number:
MDL number:
PubChem Substance ID:

assay

99%

mp

44-45 °C (lit.)

SMILES string

CCCCCCCc1ccc(cc1)C(=O)Oc2ccc(cc2)C#N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service