MilliporeSigma
  • 191418
All Photos(1)

191418

Sigma-Aldrich

1-Aminomethyl-1-cyclohexanol hydrochloride

98%

Linear Formula:
H2NCH2C6H10OH · HCl
CAS Number:
Molecular Weight:
165.66
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

217-219 °C (lit.)

SMILES string

Cl.NCC1(O)CCCCC1

InChI

1S/C7H15NO.ClH/c8-6-7(9)4-2-1-3-5-7;/h9H,1-6,8H2;1H

InChI key

DXIAEURKHWGAFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Aminomethyl-1-cyclohexanol hydrochloride was used in the synthesis of 1-aminomethyl-1-cyclohexanolarachidonylamide.

Packaging

1 g

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service