All Photos(1)

192767

Sigma-Aldrich

Ethylaluminum sesquichloride solution

25 wt. % in toluene

Linear Formula:
(C2H5)2AlCl·Cl2AlC2H5
CAS Number:
Molecular Weight:
212.05
MDL number:
PubChem Substance ID:

concentration

25 wt. % in toluene

density

0.904 g/mL at 25 °C

SMILES string

CC[Al](Cl)Cl.CC[Al](Cl)CC

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service