All Photos(1)

196762

Sigma-Aldrich

2,3,5,6-Tetrafluoroanisole

≥97%

Linear Formula:
HC6F4OCH3
CAS Number:
Molecular Weight:
180.10
EC Number:
MDL number:
PubChem Substance ID:

assay

≥97%

refractive index

n20/D 1.428 (lit.)

bp

138 °C (lit.)

density

1.293 g/mL at 25 °C (lit.)

SMILES string

COc1c(F)c(F)cc(F)c1F

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service