197459

Sigma-Aldrich

trans-1-(2-Pyridyl)-2-(4-pyridyl)-ethylene

97%

Empirical Formula (Hill Notation):
C12H10N2
CAS Number:
Molecular Weight:
182.22
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

70-72 °C (lit.)

SMILES string

[H]\C(=C(\[H])c1ccccn1)c2ccncc2

InChI

1S/C12H10N2/c1-2-8-14-12(3-1)5-4-11-6-9-13-10-7-11/h1-10H/b5-4+

InChI key

RETWYDZOYDJBIX-SNAWJCMRSA-N

Application

trans-1-(2-Pyridyl)-2-(4-pyridyl)-ethylene was used in the synthesis of new polymeric network complexes. It was also used in the preparation of a series of new ZnII and CdII supramolecular complexes.

Certificate of Analysis

Certificate of Origin

Multifarious ZnII and CdII coordination frameworks constructed by a versatile trans-1-(2-pyridyl)-2-(4-pyridyl) ethylene tecton and various benzenedicarboxyl ligands.
CrystEngComm, 12(3), 834-844 (2010)
Hierarchical regulated assembly of new metallosupramolecular networks based on metal thiocyanate and trans-1-(2-pyridyl)-2-(4-pyridyl) ethylene (bpe) via multiple interactions.
CrystEngComm, 8(7), 552-562 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service