All Photos(1)

206806

Sigma-Aldrich

Diiodofluorescein

Empirical Formula (Hill Notation):
C20H10I2O5
CAS Number:
Molecular Weight:
584.10
Colour Index Number:
45425:1
MDL number:
PubChem Substance ID:

mp

240 °C (dec.) (lit.)

λmax

522 nm

SMILES string

Oc1ccc2c(Oc3c(I)c(O)ccc3C24OC(=O)c5ccccc45)c1I

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service