Documents

210420

Sigma-Aldrich

Acid Blue 92

Dye content 40 %

Synonym(s):
Anazolene Sodium
Empirical Formula (Hill Notation):
C26H16N3Na3O10S3
CAS Number:
Molecular Weight:
695.58
Colour Index Number:
13390
EC Number:
MDL number:
PubChem Substance ID:

composition

Dye content, 40%

λmax

571 nm

SMILES string

[Na+].[Na+].[Na+].Oc1cc(cc2cc(cc(\N=N\c3ccc(Nc4ccccc4)c5c(cccc35)S([O-])(=O)=O)c12)S([O-])(=O)=O)S([O-])(=O)=O

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.