All Photos(1)

213071

Sigma-Aldrich

(−)-Carvyl propionate, mixture of isomers

98%

Empirical Formula (Hill Notation):
C13H20O2
CAS Number:
Molecular Weight:
208.30
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]20/D −107°, neat

refractive index

n20/D 1.474 (lit.)

bp

78-80 °C/0.2 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)OC1CC(CC=C1C)C(C)=C

InChI

1S/C13H20O2/c1-5-13(14)15-12-8-11(9(2)3)7-6-10(12)4/h6,11-12H,2,5,7-8H2,1,3-4H3

InChI key

DFVXNZOMAOGTBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service