All Photos(1)

213918

Sigma-Aldrich

Tungsten(VI) chloride

Synonym(s):
Tungsten hexachloride
Linear Formula:
WCl6
CAS Number:
Molecular Weight:
396.56
EC Number:
MDL number:
PubChem Substance ID:

vapor pressure

43 mmHg ( 215 °C)

bp

347 °C (lit.)

mp

275 °C (lit.)

density

3.52 g/mL at 25 °C (lit.)

SMILES string

Cl[W](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.W/h6*1H;/q;;;;;;+6/p-6

InChI key

KPGXUAIFQMJJFB-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Promotes the conversion of 1,3-dithiolanes and 1,3-dithianes to chlorinated dihydro-1,4-dithiins and dihydro-1,4-dithiepines,

Certificate of Analysis

Certificate of Origin

Firouzabadi, H. et al.
Synlett, 413-413 (1999)
Chongshen Guo et al.
Biomaterials, 52, 407-416 (2015-03-31)
Photothermal ablation (PTA) therapy has been viewed as an invasive option for cancer therapy with minimal deconstruction of healthy tissues. In this study, a potent candidate of (NH4)xWO3 nanocube was developed for PTA treatment of metastatic breast cancer in the...
Tobias Fält et al.
Journal of computer assisted tomography, 39(6), 975-980 (2015-08-22)
The aim of this study was to determine the potential of different high-Z elements to act as contrast media (CMs) alongside iodine (I) in dual-CM, dual-energy (DE) computed tomography examinations. Gadolinium (Gd), tantalum (Ta), wolfram (W), gold (Au), and bismuth...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service