All Photos(1)

217298

Sigma-Aldrich

1,3-Diallylurea

98%

Linear Formula:
H2C=CHCH2NHCONHCH2CH=CH2
CAS Number:
Molecular Weight:
140.18
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

90-93 °C (lit.)

SMILES string

C=CCNC(=O)NCC=C

InChI

1S/C7H12N2O/c1-3-5-8-7(10)9-6-4-2/h3-4H,1-2,5-6H2,(H2,8,9,10)

InChI key

QRWVOJLTHSRPOA-UHFFFAOYSA-N

Application

Intermediate for the synthesis of N-substituted heterocycles.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service