218480

Sigma-Aldrich

3-Morpholino-1,2-propanediol

≥96%

Empirical Formula (Hill Notation):
C7H15NO3
CAS Number:
Molecular Weight:
161.20
Beilstein/REAXYS Number:
107210
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

≥96%

bp

191 °C/30 mmHg (lit.)

mp

37-38 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

SMILES string

OCC(O)CN1CCOCC1

InChI

1S/C7H15NO3/c9-6-7(10)5-8-1-3-11-4-2-8/h7,9-10H,1-6H2

InChI key

VZBNUEHCOOXOHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service