MilliporeSigma
All Photos(1)

219983

Sigma-Aldrich

2-Hydroxyvaleric acid sodium salt hydrate

98%

Linear Formula:
CH3CH2CH2CH(OH)CO2Na·xH2O
CAS Number:
Molecular Weight:
140.11 (anhydrous basis)
MDL number:

assay

98%

SMILES string

O.[Na+].CCCC(O)C([O-])=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service