All Photos(1)

222046

Sigma-Aldrich

4-Butoxybenzoyl chloride

99%

Linear Formula:
CH3(CH2)3OC6H4COCl
CAS Number:
Molecular Weight:
212.67
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

99%

refractive index

n20/D 1.5495 (lit.)

bp

160 °C/8 mmHg (lit.)

density

1.122 g/mL at 25 °C (lit.)

SMILES string

CCCCOc1ccc(cc1)C(Cl)=O

InChI

1S/C11H13ClO2/c1-2-3-8-14-10-6-4-9(5-7-10)11(12)13/h4-7H,2-3,8H2,1H3

InChI key

KMGCTFHTBKBITO-UHFFFAOYSA-N

Application

4-Butoxybenzoyl chloride was used in the synthesis of 2,3-dihydro-2-(1-(4-(butoxyphenylcarbonyl)amino)-phenethyl)-1,4-dithiin-1,1,4,4-tetroxide.

Packaging

5 g in glass bottle

Certificate of Analysis

Certificate of Origin

M K Scott et al.
Bioorganic & medicinal chemistry, 8(6), 1383-1391 (2000-07-15)
The neuropeptide galanin modulates several physiological functions such as cognition, learning, feeding behavior, and depression, probably via the galanin 1 receptor (GAL-R1). Using an HTS assay based on 125I-human galanin binding to the human galanin-1 receptor (hGAL-R1), we discovered a...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service