Documents

230863

Sigma-Aldrich

Trimethylphosphine silver iodide complex

Linear Formula:
[(CH3)3P·AgI]4
CAS Number:
Molecular Weight:
1247.43
MDL number:

mp

140-142 °C (lit.)

SMILES string

[Ag]I.CP(C)C

Features and Benefits

A stable, solid, convenient source of trimethylphosphine, obtained by heating with a heat gun or an oil bath.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service