MilliporeSigma
All Photos(1)

235407

Sigma-Aldrich

(−)-N-Dodecyl-N-methylephedrinium bromide

99%

Synonym(s):
(−)-DMEB, (1R,2S)-Dodecyl(2-hydroxy-1-methyl-2-phenylethyl)dimethylammonium bromide
Linear Formula:
C6H5CH(OH)CH(CH3)N(CH3)2[(CH2)11CH3]Br
CAS Number:
Molecular Weight:
428.49
Beilstein:
5421952
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

99%

form

powder

optical activity

[α]20/D −12.5°, c = 1 in chloroform

mp

80-82 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCC[N+](C)(C)[C@@H](C)[C@H](O)c1ccccc1

InChI

1S/C23H42NO.BrH/c1-5-6-7-8-9-10-11-12-13-17-20-24(3,4)21(2)23(25)22-18-15-14-16-19-22;/h14-16,18-19,21,23,25H,5-13,17,20H2,1-4H3;1H/q+1;/p-1/t21-,23-;/m0./s1

InChI key

GOIUTZPQFMGLAH-IUQUCOCYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

5 g in glass bottle

Other Notes

Chiral phase transfer catalyst

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service