239909

Sigma-Aldrich

2-Bromopropane

≥99%

Synonym(s):
Isopropyl bromide
Linear Formula:
(CH3)2CHBr
CAS Number:
Molecular Weight:
122.99
Beilstein/REAXYS Number:
741852
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥99%

refractive index

n20/D 1.425 (lit.)

bp

59 °C (lit.)

mp

−89 °C (lit.)

solubility

alcohol: miscible(lit.)
benzene: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
water: slightly soluble(lit.)

density

1.31 g/mL at 25 °C (lit.)

SMILES string

CC(C)Br

InChI

1S/C3H7Br/c1-3(2)4/h3H,1-2H3

InChI key

NAMYKGVDVNBCFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photodissociation dynamics of 2-bromopropane was studied.

Application

2-Bromopropane (Isopropyl bromide) was used as a substitute for ozone-depleting cleaning solvents.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Supp Hazards

Repeated exposure may cause skin dryness or cracking.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

F,T

Risk Statement

60-11-48/20-66

Safety Statement

53-45

RIDADR

UN 2344 3 / PGII

WGK Germany

1

Flash Point(F)

66.2 °F

Flash Point(C)

19 °C

Rongshu Zhu et al.
The journal of physical chemistry. A, 114(21), 6188-6193 (2010-05-13)
Photodissociation dynamics of 2-bromopropane in the A band was investigated at several wavelengths between 232 and 267 nm using resonance-enhanced multiphoton ionization technique combined with velocity map ion-imaging detection. The ion images of Br ((2)P(3/2)) and Br* ((2)P(1/2)) were analyzed...
Fen Huang et al.
Wei sheng yan jiu = Journal of hygiene research, 39(1), 4-8 (2010-04-07)
To study the reproductive toxicity of male rats exposed to 1-bromopropane and 2-bromopropane and to explore the toxic mechanism of two isomers. 18 SPF SD male rats were randomly divided into 3 groups of 6 each, respectively. The groups were...
Kyung-Sun Kang et al.
Reproductive toxicology (Elmsford, N.Y.), 16(2), 151-159 (2002-04-17)
The toxic effects of 2-bromopropane (2-BP) on the reproductive tracts of male F1 offspring from dams exposed to 2-BP during gestation and lactation were investigated. Ten pregnant (sperm-positive) Sprague-Dawley rats per group were exposed sc to 2-BP at 135, 405...
H Ishikawa et al.
Reproductive toxicology (Elmsford, N.Y.), 15(1), 81-85 (2001-01-04)
We examined effects of 2-bromopropane (2-BP), a chlorofluorocarbon replacement, on mouse embryonic mutagenicity. 2-BP was administered to pregnant mice intraperitoneally (i.p.) (300, 600, 900, and 1800 mg/kg) during the early preimplantation period. On day 3 of gestation, micronuclei (MN) frequency...
S Sekiguchi et al.
Toxicology and industrial health, 16(7-8), 277-283 (2001-11-06)
The present study was performed to investigate the toxic effects of 2-bromopropane (2BP) on the female reproductive system. Female F344 rats were administered 2BP (500 or 1000 mg/kg, i.p.) at intervals of 2 or 3 days for 15-17 days. The...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.