All Photos(1)

258954

Sigma-Aldrich

1,8-Nonadiene

99%

Linear Formula:
H2C=CH(CH2)5CH=CH2
CAS Number:
Molecular Weight:
124.22
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

vapor density

>1 (vs air)

assay

99%

refractive index

n20/D 1.427 (lit.)

bp

141-142 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

C=CCCCCCC=C

InChI

1S/C9H16/c1-3-5-7-9-8-6-4-2/h3-4H,1-2,5-9H2

InChI key

VJHGSLHHMIELQD-UHFFFAOYSA-N

General description

Ring-closing metathesis reaction of 1,8-nonadiene in the presence of KCl has been studied.

Packaging

5 g in ampule

Certificate of Analysis

Certificate of Origin

Hao-Yang Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(41), 10948-10959 (2009-09-18)
Electrospray ionization mass spectrometry (ESIMS) and subsequent tandem mass spectrometry (MS/MS) analyses were used to study some important metathesis reactions with the first-generation ruthenium catalyst 1, focusing on the ruthenium complex intermediates in the catalytic cycle. In situ cationization with...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service