261181

Sigma-Aldrich

Praseodymium

foil, thickness 0.25 mm, 99.9%

Empirical Formula (Hill Notation):
Pr
CAS Number:
Molecular Weight:
140.91
EC Number:
MDL number:

assay

99.9%

form

foil

resistivity

68 μΩ-cm, 20°C

thickness

0.25 mm

bp

3520 °C (lit.)

mp

931 °C (lit.)

density

6.71 g/mL at 25 °C (lit.)

SMILES string

[Pr]

InChI

1S/Pr

InChI key

PUDIUYLPXJFUGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

hazcat

Pyr. Sol. 1

storage_class_code

4.2 - Pyrophoric and self-heating hazardous materials

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Mohamadreza K Bakht et al.
Annals of nuclear medicine, 27(3), 253-260 (2013-02-06)
Previously, a promising β(-)-emitting praseodymium-142 glass seed was proposed for brachytherapy of prostate cancer. In accordance with the previous study, a (142)Pr capillary tube-based radioactive implant (CTRI) was suggested as a source with a new structure to enhance application of...
Anupam et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 23(37), 376001-376001 (2011-09-01)
The magnetic and transport properties of PrIr(2)B(2) and PrIr(2)B(2)C have been investigated by dc and ac magnetic susceptibility, specific heat, electrical resistivity and magnetoresistance measurements. PrIr(2)B(2) forms in CaRh(2)B(2)-type orthorhombic crystal structure (space group Fddd). At low fields the dc...
Liaolin Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 223-227 (2012-04-10)
We report on the spectroscopic properties of Pr(3+)-doped boro-phosphate, boro-germo-silicate and tellurite glasses. The stimulated absorption and emission cross sections were estimated. Only one emission at 596 nm and 605 nm is observed in Pr(3+)-doped boro-phosphate and boro-germo-silicate glasses, respectively...
S Majumdar et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(6), 066005-066005 (2013-01-12)
We report on the effect of oxygen on the Jahn-Teller distortion and dynamic magnetic properties of low hole-doped Pr(0.9)Ca(0.1)MnO(3) thin films, using micro-Raman spectroscopy and the temperature dependent ac susceptibility, as a function of the frequency, dc field bias, and...
D V Sunitha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 279-287 (2012-10-23)
A series of Pr(3+) (1-9 mol%) doped CdSiO(3) nanophosphors have been prepared for the first time by a low temperature solution combustion method using oxalyldihydrizide (ODH) as a fuel. The final product was characterized by Powder X-ray diffraction (PXRD), Fourier...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service