MilliporeSigma
All Photos(1)

261688

Sigma-Aldrich

Methyldiphenylsilane

≥97%

Synonym(s):
Diphenylmethylsilane
Linear Formula:
(C6H5)2SiHCH3
CAS Number:
Molecular Weight:
198.34
Beilstein:
1867183
EC Number:
MDL number:
PubChem Substance ID:

assay

≥97%

refractive index

n20/D 1.571 (lit.)

bp

93-94 °C/1 mmHg (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

C[SiH](c1ccccc1)c2ccccc2

InChI

1S/C13H14Si/c1-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11,14H,1H3

InChI key

FULSRCPEOUATID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service