265616

Sigma-Aldrich

Silver

wire, diam. 1.5 mm, 99.9% trace metals basis

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
PubChem Substance ID:

assay

99.9% trace metals basis

form

wire

resistivity

1.59 μΩ-cm, 20°C

diam.

1.5 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Quantity

10 g = 50 cm

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

hazcat

Aquatic Acute 1 - Aquatic Chronic 1

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Sa Ram Lee et al.
Journal of biomedical nanotechnology, 9(7), 1241-1244 (2013-08-06)
We demonstrate simultaneous detection of surface-enhanced Raman scattering (SERS) and fluorescence signals from a silver microbead. For the dual signal generation, silver microbeads with a diameter of 15 microm were functionalized with benzenethiol (BT) as a Raman tag and a...
Yumi Kim et al.
Journal of nanoscience and nanotechnology, 13(6), 3897-3900 (2013-07-19)
The objectives of this research were to study biogenic synthesis of Ag(0) nanoparticles and to determine the effects of various experimental conditions such as silver nitrate concentrations, pHs and temperatures controlling for the optimal biosynthesis of Ag(0) nanoparticles. The metal-reducing...
R Nirmala et al.
Journal of nanoscience and nanotechnology, 13(7), 4686-4693 (2013-08-02)
We report on the preparation and characterization of polyurethane (PU) nanofibers containing silver (Ag) nanoparticles were synthesized by using electrospinning. Two different approaches were adopted to incorporate the Ag nanoparticles in to PU nanofibers. In the first approach, a homogeneous...
Jaya Sundaram et al.
Journal of nanoscience and nanotechnology, 13(8), 5382-5390 (2013-07-26)
Silver/biopolymer nanoparticles were prepared by adding 100 mg silver nitrate to 2% polyvinyl alcohol solution and reduced the silver nitrate using 2% trisodium citrate for high performance Surface Enhanced Raman Scattering (SERS) substrates. Optical properties of nanoparticle were measured using...
T Prameela Devi et al.
Indian journal of experimental biology, 51(7), 543-547 (2013-08-01)
A total of 75 isolates belonging to five different species of Trichoderma viz., T. asperellum, T. harzianum, T. longibrachiatum, T. pseudokoningii and T. virens were screened for the production of silver nanoparticles. Although all the isolates produced nanoparticles, T. virens...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service