MilliporeSigma
All Photos(1)

269077

Sigma-Aldrich

Indole

98%

Synonym(s):
1H-Benzo[b]pyrrole
Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
Beilstein:
107693
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

bp

253-254 °C (lit.)

mp

51-54 °C (lit.)

SMILES string

c1ccc2[nH]ccc2c1

InChI

1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H

InChI key

SIKJAQJRHWYJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service