MilliporeSigma
All Photos(1)

278726

Sigma-Aldrich

Tetraammineplatinum(II) nitrate

≥50.0% Pt basis

Linear Formula:
[Pt(NH3)4](NO3)2
CAS Number:
Molecular Weight:
387.21
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥50.0% Pt basis

form

solid

reaction suitability

core: platinum
reagent type: catalyst

mp

262 °C (dec.) (lit.)

SMILES string

N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.4H3N.Pt/c2*2-1(3)4;;;;;/h;;4*1H3;/q2*-1;;;;;+2

InChI key

RBAKORNXYLGSJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetraammineplatinum(II) nitrate can be used as a platinum source to prepare:
  • Pt/C catalyst via incipient-wetness impregnation technique.
  • Platinum loaded zeolite catalyst with acidic and oxidizing properties for the oxidation of different chlorinated hydrocarbons.
  • Bimetallic Pt/Zn catalyst for the selective hydrogenation of crotonaldehyde.

Packaging

1, 5 g in glass bottle

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service