All Photos(1)

283487

Sigma-Aldrich

3,5-Diiodoanthranilic acid

99%

Synonym(s):
2-Amino-3,5-diiodobenzoic acid
Linear Formula:
I2C6H2(NH2)CO2H
CAS Number:
Molecular Weight:
388.93
EC Number:
MDL number:
PubChem Substance ID:

assay

99%

mp

241-243 °C (lit.)

SMILES string

Nc1c(I)cc(I)cc1C(O)=O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service