MilliporeSigma
All Photos(1)

287563

Sigma-Aldrich

Bis(2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl) camphorate

technical grade, 85%

Empirical Formula (Hill Notation):
C24H20F24O4
CAS Number:
Molecular Weight:
828.37
MDL number:
PubChem Substance ID:

grade

technical grade

assay

85%

refractive index

n20/D 1.372 (lit.)

bp

174-177 °C/0.8 mmHg (lit.)

density

1.624 g/mL at 25 °C (lit.)

SMILES string

CC1(C)C(CCC1(C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service