MilliporeSigma
All Photos(1)

301906

Sigma-Aldrich

8-Hydroxy-7-(4-sulfo-1-naphthylazo)-5-quinolinesulfonic acid disodium salt

Synonym(s):
SNAZOXS
Empirical Formula (Hill Notation):
C19H11N3Na2O7S2
CAS Number:
Molecular Weight:
503.42
EC Number:
MDL number:
PubChem Substance ID:

SMILES string

[Na+].[Na+].Oc1c(cc(c2cccnc12)S([O-])(=O)=O)N=Nc3ccc(c4ccccc34)S([O-])(=O)=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service