MilliporeSigma
All Photos(1)

306398

Sigma-Aldrich

α,α,α,2,3,5,6-Heptafluorotoluene

99%

Synonym(s):
2,3,5,6-Tetrafluorobenzotrifluoride
Linear Formula:
HC6F4CF3
CAS Number:
Molecular Weight:
218.07
MDL number:

assay

99%

refractive index

n20/D 1.379 (lit.)

bp

111-112 °C (lit.)

density

1.601 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(F)c(F)c(c1F)C(F)(F)F

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service