MilliporeSigma
All Photos(1)

306428

Sigma-Aldrich

(+)-3,9-Dibromocamphor

98%

Synonym(s):
3,9-Dibromo-(+)-camphor
Empirical Formula (Hill Notation):
C10H14Br2O
CAS Number:
Molecular Weight:
310.03
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]19/D +100°, c = 1 in chloroform

mp

156-159 °C (lit.)

SMILES string

[H]C1(Br)C2CCC(C)(C1=O)C2(C)CBr

InChI

1S/C10H14Br2O/c1-9-4-3-6(7(12)8(9)13)10(9,2)5-11/h6-7H,3-5H2,1-2H3/t6-,7+,9+,10?/m1/s1

InChI key

DCDNKSJBRIJYEC-MZIHZIJQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service