MilliporeSigma
All Photos(1)

345903

Sigma-Aldrich

4,6-Diphenylthieno[3,4-d]-1,3-dioxol-2-one 5,5-dioxide

95%

Synonym(s):
Steglich’s reagent, TDO-carbonate
Empirical Formula (Hill Notation):
C17H10O5S
CAS Number:
Molecular Weight:
326.32
Beilstein:
1396884
EC Number:
MDL number:
PubChem Substance ID:

assay

95%

mp

230-290 °C (lit.)

SMILES string

O=C1OC2=C(c3ccccc3)S(=O)(=O)C(=C2O1)c4ccccc4

InChI

1S/C17H10O5S/c18-17-21-13-14(22-17)16(12-9-5-2-6-10-12)23(19,20)15(13)11-7-3-1-4-8-11/h1-10H

InChI key

UEOYFCIGZWQXTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service