All Photos(1)

349577

Sigma-Aldrich

2-Acetyl-2-thiazoline

99%

Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18
Beilstein:
1209707
MDL number:
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.5305 (lit.)

mp

26-30 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C1=NCCS1

InChI

1S/C5H7NOS/c1-4(7)5-6-2-3-8-5/h2-3H2,1H3

InChI key

FZOZFDAMVVEZSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Kenji Kumazawa et al.
Journal of agricultural and food chemistry, 50(20), 5660-5663 (2002-09-19)
Two kinds of pan-fired green teas (Japanese Kamairi-cha and Chinese Longing tea) were compared with the common Japanese green tea (Sen-cha). Application of the aroma extract dilution analysis (AEDA) using the volatile fraction of the Sen-cha, Kamairi-cha and Longing tea...
Chemistry of 2-acetyl-1-pyrroline, 6-acetyl-1,2,3,4-tetrahydropyridine, 2-acetyl-2-thiazoline, and 5-acetyl-2,3-dihydro-4H-thiazine: extraordinary Maillard flavor compounds.
An Adams et al.
Chemical reviews, 106(6), 2299-2319 (2006-06-15)
Rachid Bel Rhlid et al.
Journal of agricultural and food chemistry, 50(8), 2350-2355 (2002-04-04)
Roasted notes contribute to the flavor of thermally processed foods such as meat and bread. 2-Acetyl-2-thiazoline is one of the key volatile compounds responsible for the roasted and popcorn-like aroma character. We report here on the biogeneration of flavoring preparations...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service