356840

Sigma-Aldrich

Iron

wire, diam. 0.25 mm, ≥99.9%

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:

assay

≥99.9%

form

wire

resistivity

9.71 μΩ-cm

diam.

0.25 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service