357030

Sigma-Aldrich

Cobalt

wire, diam. 0.5 mm, ≥99.9%

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥99.9%

form

wire

resistivity

6.24 μΩ-cm, 20°C

diam.

0.5 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

D W Su et al.
Journal of nanoscience and nanotechnology, 13(5), 3354-3359 (2013-07-19)
Highly ordered mesoporous Co3O4 nanostructures were prepared using SBA-15 silica as hard templates. The mesoporous structures were characterized by X-ray diffraction, high resolution transmission electron microscopy, and N2 adsorption/desorption isotherm analysis. The results demonstrated that the as-prepared mesoporous Co3O4 has...
M K Shobana et al.
Journal of nanoscience and nanotechnology, 13(5), 3535-3538 (2013-07-19)
Ferrites are extremely important magnetic ceramics in the production of electronic components because they reduce the energy losses by the induced currents acting as electrical insulators. Similarly, the spinel-structured cobalt-based ferrites are promising materials for stress, torsion sensors and energy...
Heather De Bari et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 69(Pt 3), 228-236 (2013-03-23)
The X-ray crystal structure of ribosome hibernation promoting factor (HPF) from Vibrio cholerae is presented at 2.0 Å resolution. The crystal was phased by two-wavelength MAD using cocrystallized cobalt. The asymmetric unit contained two molecules of HPF linked by four Co...
S Perconti et al.
Journal of biological regulators and homeostatic agents, 27(2), 443-454 (2013-07-09)
Size-dependent characteristics of novel engineered nanomaterials might result in unforeseen biological responses and toxicity. To address this issue, we used cDNA microarray analysis (13443 genes) coupled with bioinformatics and functional gene annotation studies to investigate the transcriptional profiles of Balb/3T3...
A J Hart et al.
The Journal of bone and joint surgery. American volume, 95(8), 678-685 (2013-04-19)
Determining the relationship between clinical factors and engineering analysis of retrieved hip implants can help our understanding of the mechanism of device failure. This is particularly important for metal-on-metal hip arthroplasties because the most common cause of failure is unexplained....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service