357138

Sigma-Aldrich

Rhenium

wire, diam. 1.0 mm, ≥99.9% trace metals basis

Synonym(s):
Rhenium element
Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥99.9% trace metals basis

form

wire

diam.

1.0 mm

bp

5596 °C (lit.)
5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

SMILES string

ReH

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

1.6 g = 100 mm

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

F

Risk Statement

11

Safety Statement

16

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Raphael Horvath et al.
Inorganic chemistry, 52(3), 1304-1317 (2013-01-15)
Transition-metal complexes of the types [Re(CO)(3)Cl(NN)], [Re(CO)(3)py(NN)](+), and [Cu(PPh(3))(2)(NN)](+), where NN = 4,4'-bis(5-phenyl-1,3,4-oxadiazol-2-yl)-2,2'-bipyridine (OX) and 4,4'-bis(N,N-diphenyl-4-[ethen-1-yl]-aniline)-2,2'-bipyridine (DPA), have been synthesized and characterized. Crystal structures for [Re(CO)(3)Cl(DPA)] and [Cu(PPh(3))(2)(OX)]BF(4) are presented. The crystal structure of the rhenium complex shows a trans...
Wilber Quispe-Tintaya et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(21), 8668-8673 (2013-04-24)
No significant improvement in therapy of pancreatic cancer has been reported over the last 25 y, underscoring the urgent need for new alternative therapies. Here, we coupled a radioisotope, (188)Rhenium, to an attenuated (at) live Listeria monocytogenes (Listeria(at)) using Listeria-binding...
Ties J Korstanje et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(39), 13224-13234 (2013-08-16)
Rhenium-based complexes are powerful catalysts for the dehydration of various alcohols to the corresponding olefins. Here, we report on both experimental and theoretical (DFT) studies into the mechanism of the rhenium-catalyzed dehydration of alcohols to olefins in general, and the...
Titel Jurca et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4278-4286 (2013-02-02)
The development of rhenium(I) chemistry has been restricted by the limited structural and electronic variability of the common pseudo-octahedral products fac-[ReX(CO)3L2] (L2 = α-diimine). We address this constraint by first preparing the bidentate bis(imino)pyridine complexes [(2,6-{2,6-Me2C6H3N=CPh}2C5H3N)Re(CO)3X] (X = Cl 2...
Jonathan M Smieja et al.
Inorganic chemistry, 52(5), 2484-2491 (2013-02-20)
Electrocatalytic properties, X-ray crystallographic studies, and infrared spectroelectrochemistry (IR-SEC) of Mn(bpy-tBu)(CO)3Br and [Mn(bpy-tBu)(CO)3(MeCN)](OTf) are reported. Addition of Brönsted acids to CO2-saturated solutions of these Mn complexes and subsequent reduction of the complexes lead to the stable and efficient production of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.