376671

Sigma-Aldrich

Diisoheptyl phthalate

ester content ≥99 % (mixture of C7 isomers), technical grade

Linear Formula:
C6H4-1,2-(CO2C7H15)2
CAS Number:
Molecular Weight:
362.50
EC Number:
PubChem Substance ID:

grade

technical grade

vapor pressure

<5 mmHg ( 20 °C)

composition

ester content, ≥99% (mixture of C7 isomers)

impurities

≤0.6% dioctyl phthalate

refractive index

n20/D 1.486 (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCOC(=O)c1ccccc1C(=O)OCCCCC(C)C

InChI

1S/C22H34O4/c1-17(2)11-7-9-15-25-21(23)19-13-5-6-14-20(19)22(24)26-16-10-8-12-18(3)4/h5-6,13-14,17-18H,7-12,15-16H2,1-4H3

InChI key

RKELNIPLHQEBJO-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Justin M Conley et al.
Toxicological sciences : an official journal of the Society of Toxicology, 164(1), 166-178 (2018-06-27)
Biomonitoring efforts have clearly shown that all humans are exposed to chemical mixtures. Of concern is whether or not exposure to mixtures during pregnancy contributes to congenital abnormalities in children even when each chemical is at an individual dose that...
Soy Kim et al.
Marine pollution bulletin, 151, 110824-110824 (2020-02-15)
Due to strong regulation of phthalates, non-phthalate plasticizers (NPPs) have been introduced to the industrial market. In this study, concentrations of 16 phthalates and five NPPs in sediment were measured to investigate the occurrence, potential source, and ecotoxicological implications of...
Young-Sun Lee et al.
Marine pollution bulletin, 154, 111119-111119 (2020-04-23)
Due to regulations on phthalates, non-phthalate plasticizers (NPPs) are now used as an alternative. Limited studies have been conducted on the occurrence and distribution of NPPs. In this study, sediment samples were collected from 50 locations along the Korean coast...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service