MilliporeSigma
All Photos(1)

378909

Sigma-Aldrich

Ethyl 3-hydrazino-3-oxopropionate

97%

Synonym(s):
Ethyl malonyl hydrazide
Linear Formula:
H2NNHCOCH2CO2C2H5
CAS Number:
Molecular Weight:
146.14
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

form

solid

mp

72-75 °C (lit.)

SMILES string

CCOC(=O)CC(=O)NN

InChI

1S/C5H10N2O3/c1-2-10-5(9)3-4(8)7-6/h2-3,6H2,1H3,(H,7,8)

InChI key

HCPOCMMGKBZWSJ-UHFFFAOYSA-N

Related Categories

Application

Ethyl 3-hydrazino-3-oxopropionate may be used in the following studies:
  • As starting reagent in the step-wise construction of complex pyrrole-pyrazole system.,
  • Synthesis of 4,5-dihydro-1H-pyrazoles, via reaction with 4-alkoxy-1,1,1-trifluoro-3-alken-2-ones, using conventional thermal heating method and a microwave-assisted method.
  • Synthesis of pyrazolopyridazine.

Packaging

25 g in glass bottle

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service