All Photos(1)

381497

Sigma-Aldrich

Trimethylolpropane tris(2-mercaptoacetate)

technical grade

Linear Formula:
(HSCH2CO2CH2)3CC2H5
CAS Number:
Molecular Weight:
356.48
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

refractive index

n20/D 1.529 (lit.)

bp

245 °C/1 mmHg (lit.)

density

1.28 g/mL at 25 °C (lit.)

SMILES string

CCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS

InChI

1S/C12H20O6S3/c1-2-12(6-16-9(13)3-19,7-17-10(14)4-20)8-18-11(15)5-21/h19-21H,2-8H2,1H3

InChI key

KAJBSGLXSREIHP-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service