388602

Sigma-Aldrich

2-Heptyne

98%

Synonym(s):
Butylmethylacetylene
Linear Formula:
CH3(CH2)3C≡CCH3
CAS Number:
Molecular Weight:
96.17
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98%

refractive index

n20/D 1.423 (lit.)

bp

110-111 °C/747 mmHg (lit.)

density

0.745 g/mL at 25 °C (lit.)

SMILES string

CCCCC#CC

InChI

1S/C7H12/c1-3-5-7-6-4-2/h3,5,7H2,1-2H3

InChI key

AMSFEMSYKQQCHL-UHFFFAOYSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

F,Xi

Risk Statement

11-36/37/38

Safety Statement

16-26

RIDADR

UN 3295BE 3 / PGII

WGK Germany

3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.