401536

Sigma-Aldrich

Copper(II) fluoride hydrate

99.999% trace metals basis

Empirical Formula (Hill Notation):
CuF2 · xH2O
Molecular Weight:
101.54 (anhydrous basis)
EC Number:
MDL number:
PubChem Substance ID:

assay

99.999% trace metals basis

form

powder

density

2.93 g/mL at 25 °C (lit.)

Featured Industry

Battery Manufacturing

SMILES string

O.F[Cu]F

InChI

1S/Cu.2FH.H2O/h;2*1H;1H2/q+2;;;/p-2

InChI key

YZWULHCOBMVFKP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3260 8 / PGIII

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.