All Photos(1)

404918

Sigma-Aldrich

2-Chloroethyl carbamate

97%

Linear Formula:
H2NCO2CH2CH2Cl
CAS Number:
Molecular Weight:
123.54
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

74-77 °C (lit.)

SMILES string

NC(=O)OCCCl

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service