All Photos(1)

409278

Sigma-Aldrich

Zinc iodide

anhydrous, beads, −10 mesh, 99.999% trace metals basis

Synonym(s):
Diiodozinc, Zinc diiodide
Linear Formula:
ZnI2
CAS Number:
Molecular Weight:
319.20
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.23

Quality Level

grade

anhydrous

assay

99.999% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: zinc

particle size

−10 mesh

mp

445 °C (lit.)

density

4.74 g/mL at 25 °C (lit.)

SMILES string

I[Zn]I

InChI

1S/2HI.Zn/h2*1H;/q;;+2/p-2

InChI key

UAYWVJHJZHQCIE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK Germany

WGK 3

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Y P Chau et al.
Histology and histopathology, 9(4), 649-656 (1994-10-01)
Cytochemical relationship between Golgi complex and dense-cored granules (DCGs) of small granule-containing (SGC) cells in rat superior cervical ganglia was examined in electron microscopy by zinc-iodide-osmium tetroxide (ZIO) method and by enzyme cytochemistry for thiamine pyrophosphatase (TPPase) and acid phosphatase...
Yuko Nakamura et al.
Journal of endodontics, 31(10), 755-758 (2005-09-28)
The purpose of this study was to investigate the cytotoxicity of iontophoresis treatment using direct current (DC) with or without antibacterial agents. The following antibacterial agents were used: diamine silver fluoride (AgF); sodium fluoride (NaF); and iodine zinc iodide (JJZ)....
T Müller
Histology and histopathology, 15(2), 493-498 (2000-05-16)
The nerve fiber distribution in the epidermis of the hairless rat skin was studied light microscopically by means of zinc iodide-osmium tetroxide staining. Two different morphological types of free nerve fiber endings could be detected: clusters of relatively thick nerve...
Mohammad Saquib et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(24), 6041-6049 (2009-05-07)
Simple and efficient syntheses, catalysed by a mixed Lewis acid system (ZrCl(4)/ZnI(2)), of enantiomerically pure 2- and 2,3-disubstituted furan derivatives--including important synthons such as 3-iodofuran and 3-(hydroxymethyl)furan derivatives--from commercially available 3,4,6-tri-O-acetyl-D-glucal are described. The transformation is achieved through a synergistic...
Lei Han et al.
Inorganic chemistry, 46(5), 1511-1513 (2007-02-08)
Hydrothermal reaction of 4,4-trimethylenedipyridine (tmdp) with ZnI2 under 175 degrees C yields a novel compound, {[Zn2I4(tmdp)2]n.[Zn2I4(tmdp)2]n}, which has a chiral infinite double-stranded helical structure consisting of two single-stranded helices of the same handedness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service