MilliporeSigma
All Photos(1)

410993

Sigma-Aldrich

(−)-DIP-Bromide

95%

Synonym(s):
(1R)-(−)-B-Bromodiisopinocampheylborane
Empirical Formula (Hill Notation):
C20H34BBr
CAS Number:
Molecular Weight:
365.20
MDL number:
PubChem Substance ID:

assay

95%

SMILES string

C[C@H]1[C@@H](CC2CC1C2(C)C)B(Br)[C@@H]3CC4CC([C@H]3C)C4(C)C

Application

Both enantiomers have been used for asymmetric reduction of prochiral ketones, enolboration reactions, and opening of meso-epoxides to form non-racemic halohydrins.

Legal Information

DIP-Bromide is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service